Learn More
Invitrogen™ IRF5 Recombinant Superclonal™ Antibody (7 HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711293
Description
This antibody is predicted to react with Monkey, Cat, Pig and Dog Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
Specifications
| IRF5 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q13568 | |
| IRF5 | |
| Peptides corresponding to Human IRF5 (aa 402-417, 129-143). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Immunocytochemistry | |
| 7 HCLC | |
| Unconjugated | |
| IRF5 | |
| AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.