missing translation for 'onlineSavingsMsg'
Learn More

IRF5 Rabbit anti-Human, Mouse, Clone: 6L2B5, Novus Biologicals™

Product Code. 18320443
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320443 20 μg 20µL
18300336 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18320443 Supplier Bio-Techne Supplier No. NBP31534320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IRF5 Monoclonal antibody specifically detects IRF5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF5
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 6L2B5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 6-151 of human IRF5 (Q13568). PVAPTPPRRVRLKPWLVAQVNSCQYPGLQWVNGEKKLFCIPWRHATRHGPSQDGDNTIFKAWAKETGKYTEGVDEADPAKWKANLRCALNKSRDFRLIYDGPRDMPPQPYKIYEVCSNGPAPTDSQPPEDYSFGAGEEEEEEEELQ
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3663
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.