missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Rabbit anti-Human, Mouse, Clone: 6L2B5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15343-100UL
This item is not returnable.
View return policy
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| IRF5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 6L2B5 | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200 | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 6-151 of human IRF5 (Q13568). PVAPTPPRRVRLKPWLVAQVNSCQYPGLQWVNGEKKLFCIPWRHATRHGPSQDGDNTIFKAWAKETGKYTEGVDEADPAKWKANLRCALNKSRDFRLIYDGPRDMPPQPYKIYEVCSNGPAPTDSQPPEDYSFGAGEEEEEEEELQ | |
| 100 μg | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction