missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF5 Monoclonal Antibody (K40049_2A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555782
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TGKYTEGVDE ADPAKWKANL RCALNKSRDF RLIYDGPRDM PPQPYKIYEV CSNGPAPTDS QPPEDYSFGA GEEEEEEEEL QRMLPSLSLT EDVKWPPTLQ
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
Specifications
| IRF5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13568 | |
| IRF5 | |
| Recombinant human IRF5 (immunogen range is 71-170). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K40049_2A6 | |
| Unconjugated | |
| IRF5 | |
| AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering