missing translation for 'onlineSavingsMsg'
Learn More

IRF5 Antibody (10T1), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18412748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412748 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412748 Supplier Novus Biologicals Supplier No. NBP104307MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF5 Monoclonal antibody specifically detects IRF5 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF5
Applications Flow Cytometry
Classification Monoclonal
Clone 10T1
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Host Species Mouse
Immunogen Recombinant human IRF5 (176-240 aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3663
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.