missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Antibody (CL14615) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26863MFV450
This item is not returnable.
View return policy
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| IRF4 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| This antibody was developed using a recombinant protein derived from Q15306, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 3662 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| CL14615 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction