missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Antibody (CL14615) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26863R
This item is not returnable.
View return policy
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| IRF4 | |
| Monoclonal | |
| DyLight 550 | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| This antibody was developed using a recombinant protein derived from Q15306, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 3662 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| CL14615 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur