missing translation for 'onlineSavingsMsg'
Learn More

IRF4 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18407867 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407867 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407867 Supplier Novus Biologicals Supplier No. NBP176574AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRF4 Polyclonal antibody specifically detects IRF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human IRF4. The immunogen is located within the last 50 amino acids of IRF4. Amino Acid Squence: NPEDYHRSIRHSSIQE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3662
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.