missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3, p Ser386 Antibody (SR1819), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21582-100ul
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| IRF3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1819 | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| interferon regulatory factor 3, IRF-3 | |
| A synthesized peptide derived from human phospho-IRF3 [Ser386] (Uniprot #: Q14653) | |
| 100 μg | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction