missing translation for 'onlineSavingsMsg'
Learn More

IRF3 Mouse anti-Human, Clone: PCRP-IRF3-3B2, Novus Biologicals™

Product Code. 18320882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320882 100 μg 100µg
18346784 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18320882 Supplier Novus Biologicals Supplier No. NBP313954100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF3
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-IRF3-3B2
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry, Immunofluorescence 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q14653
Gene Alias interferon regulatory factor 3, IRF-3
Host Species Mouse
Immunogen Recombinant full-length human IRF3 protein (Uniprot: Q14653)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3661
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.