missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (PCRP-IRF3-3B2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14106H
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot
Specifications
| IRF3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| PCRP-IRF3-3B2 | |
| Western Blot | |
| interferon regulatory factor 3, IRF-3 | |
| Recombinant full-length human IRF3 protein (Uniprot: Q14653) | |
| 0.1 mL | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction