missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£232.00 - £419.00
Specifications
| Antigen | IRF3 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491922
|
Novus Biologicals
NBP1-92028-25ul |
25 μL |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18484182
|
Novus Biologicals
NBP1-92028 |
0.1 mL |
£419.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRF3 Polyclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| IRF3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| PBS (pH 7.2) and 40% Glycerol | |
| 3661 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| interferon regulatory factor 3, IRF-3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VNKSRTRFRIPWKHGLRQDAQQEDFGIFQAWAEATGAYVPGRDKPDLPTWKRNFRSALNRKEGLRLAEDRSKDPHDPHKIYEFVNSGVGDFSQPDTSPDTNGGGSTSDTQEDILDELLGNMVLAPLPDPGPPSLAVAPEPCPQ | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title