missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76598
This item is not returnable.
View return policy
Description
IRF3 Polyclonal specifically detects IRF3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| IRF3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| interferon regulatory factor 3, IRF-3 | |
| Rabbit | |
| 50 kDa | |
| 0.1 mg | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| NP_001562 | |
| IRF3 | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the center of human IRF3. The immunogen is located within amino acids 150 - 200 of IRF3. Amino Acid Squence: CPQPLRSPSLDNPT | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction