missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (3F10), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04308AF532
This item is not returnable.
View return policy
Description
IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| IRF3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IRF3 | |
| Recombinant human IRF-3(108-166aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| 3F10 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| interferon regulatory factor 3, IRF-3 | |
| Mouse | |
| Protein G purified | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction