missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IRF2BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28933
This item is not returnable.
View return policy
Description
IRF2BP2 Polyclonal antibody specifically detects IRF2BP2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| IRF2BP2 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 359948 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| interferon regulatory factor 2 binding protein 2, IRF-2-binding protein 2 | |
| between 450 and 500 | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction