missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF2BP2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81996F
This item is not returnable.
View return policy
Description
IRF2BP2 Polyclonal antibody specifically detects IRF2BP2 in Human, Mouse samples. It is validated for ELISA
Specifications
| IRF2BP2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 359948 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| FITC | |
| interferon regulatory factor 2 binding protein 2, IRF-2-binding protein 2 | |
| IRF2BP2 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human IRF2BP2. The immunogen is located within amino acids 440 - 490 of IRF2BP2. Amino Acid Squence: HSTTRRNSNSPPSPSSMN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction