missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF2BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559898
This item is not returnable.
View return policy
Description
Immunogen sequence: LGSMPGLMPP GLLAAAVSGL GSRGLTLAPG LSPARPLFGS DFEKEKQQRN ADCLAELNEA MRGRAEEWHG RPKAVREQL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.
Specifications
| IRF2BP1 | |
| Polyclonal | |
| Unconjugated | |
| Irf2bp1 | |
| 6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IU81 | |
| Irf2bp1 | |
| Recombinant protein corresponding to Human IRF2BP1. Recombinant protein control fragment (Product #RP-99170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu