missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF2BP1 Monoclonal Antibody (OTI3A6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527521
This item is not returnable.
View return policy
Description
IRF2BP1 Monoclonal Antibody for Western Blot, IHC (P)
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.
Specifications
| IRF2BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IU81 | |
| Irf2bp1 | |
| Human recombinant protein fragment corresponding to amino acids 457-584 of IRF2BP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A6 | |
| Unconjugated | |
| Irf2bp1 | |
| 6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26145 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction