missing translation for 'onlineSavingsMsg'
Learn More

IRF2BP1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30737630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737630 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737630 Supplier Novus Biologicals Supplier No. AF6800Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IRF2BP1 Polyclonal antibody specifically detects IRF2BP1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF2BP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias DKFZP434M154, interferon regulatory factor 2 binding protein 1, interferon regulatory factor 2-binding protein 1, IRF-2-binding protein 1, IRF-2BP1
Host Species Sheep
Immunogen E. coli-derived recombinant human IRF2BP1, Ala188-Glu356, Accession # Q8IU81
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26145
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.