missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518083
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
Specifications
| IRF2 | |
| Polyclonal | |
| Unconjugated | |
| IRF2 | |
| 9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3660 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P14316 | |
| IRF2 | |
| Synthetic peptide sequence (KTSDITQARVKSC) corresponding to the C-terminus amino acids of IRF2 (aa 337-349). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction