missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584335
This item is not returnable.
View return policy
Description
Immunogen sequence: SMSELYPLQI SPVSSYAESE TTDSVPSDEE SAEGRPHWRK RNIEGKQYLS NMGTRGSYLL PGMASFVTSN KPDLQVTIKE ESNPVPYNSS WPPFQDLPLS SSMTPASSSS RPDRETRASV IKKTSD Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.
IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
Specifications
| IRF2 | |
| Polyclonal | |
| Unconjugated | |
| IRF2 | |
| 9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14316 | |
| IRF2 | |
| Recombinant protein corresponding to Human IRF2. Recombinant protein control fragment (Product #RP-105066). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction