missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148862
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
Specifications
| IRF2 | |
| Polyclonal | |
| Unconjugated | |
| IRF2 | |
| 9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 16363, 3660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P14316, P23906 | |
| IRF2 | |
| E. coli-derived recombinant mouse IRF2, Val151-Cys349. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction