missing translation for 'onlineSavingsMsg'
Learn More

IRF1 Antibody (BLR039F), Novus Biologicals™

Product Code. 18345190 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345190 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345190 Supplier Novus Biologicals Supplier No. NBP276419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IRF1 Monoclonal specifically detects IRF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone BLR039F
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 20 μL/1mg lysate, Immunocytochemistry 1:100-1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide
Gene Alias interferon regulatory factor 1, MARIRF-1
Gene Symbols IRF1
Host Species Rabbit
Immunogen Immunogen sequence between amino acids 250 and 300
Purification Method Antigen Affinity-purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3659.0
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.