missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IREB2 Recombinant Rabbit Monoclonal Antibody (1Q7R2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558032

 View more versions of this product

Product Code. 30283327

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SLEAPDSVLF PWDLKSTYIR CPSFFDKLTK EPIALQAIEN AHVLLYLGDS VTTDHISPAG SIARNSAAAK YLTNRGLTPR EFNSYGARRG NDAVMTRGTF ANIKLFNKFI GKPAPKTIHF PSGQTLDVFE AAELYQKEGI PLIILAGKKY GSGNSRDWAA KGPYLLGVKA VLAESYEKIH KDHLIGIGIA PLQFLPGENA DSLGLSGRET FSLTFPEELS PGITLNIQTS TGKVFSVIAS FEDDVEITLY KHGGLLNFVA RKFS.

Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IREB2
Recombinant Monoclonal
0.25 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P48200, Q62751, Q811J3
IREB2
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
1Q7R2
Unconjugated
IREB2
ACO3; D9Ertd85e; FLJ23381; im:7153062; Ireb2; IRE-BP 2; IREBP2; Iron regulatory protein 2; iron responsive element binding protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2; IRP2AD
Rabbit
Protein A
RUO
3658, 64602, 64831
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.