missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IREB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564388
This item is not returnable.
View return policy
Description
Immunogen sequence: EYGAILSFFP VDNVTLKHLE HTGFSKAKLE SMETYLKAVK LFRNDQNSSG EPEYSQVIQI NLNSIVPSVS GP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
Specifications
| IREB2 | |
| Polyclonal | |
| Unconjugated | |
| IREB2 | |
| ACO3; D9Ertd85e; FLJ23381; im:7153062; Ireb2; IRE-BP 2; IREBP2; Iron regulatory protein 2; iron responsive element binding protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2; IRP2AD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48200 | |
| IREB2 | |
| Recombinant protein corresponding to Human IREB2. Recombinant protein control fragment (Product #RP-104254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction