missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1 alpha, p Ser724 Antibody (1F12), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £406.00
Specifications
| Antigen | IRE1 alpha |
|---|---|
| Clone | 1F12 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038407
|
Novus Biologicals
NBP3-26425-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029902
|
Novus Biologicals
NBP3-26425-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRE1 alpha Monoclonal antibody specifically detects IRE1 alpha in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
| IRE1 alpha | |
| Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| A synthesized peptide derived from Human IRE1 alpha [p Ser724] [UniProt O75460] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1F12 | |
| Western Blot, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Diabetes Research, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction, Unfolded Protein Response | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2081 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title