missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IRDye™ 800CW, NHS ester

Product Code. 30306103
Click to view available options
:
500 μg
This item is not returnable. View return policy

Product Code. 30306103

Brand: Tocris Bioscience™ 8881/500U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Near-infared dye (reactive group NHS ester)

Key information: IRDye™ 800CW, NHS ester is a near-infrared (NIR) fluorescent dye, supplied with an NHS ester reactive group for the labeling of primary amines.

Application: Fluorescence microscopy.

Properties and Photophysical Data: Excitation and emission maxima (λ) are 773 nm and 792 nm, respectively.

Please see the protocol for further information on protein/antibody labeling and conjugation.

It is recommended to dissolve this dye in water or DMSO (10 - 20 mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2-((E)-2-((E)-3-(2-((E)-1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)ethylidene)-2-(4-sulfonatophenoxy)cyclohex-1-en-1-yl)vinyl)-3,3-dimethyl-1-(4-sulfonatobutyl)-3H-indol-1-ium-5-sulfonate
CAS 956579-01-4
Quantity 500 μg
Target Fluorescent Dyes
Molecular Formula C50H54N3Na3O17S4
Purity 0.8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis