missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IRDye™ 800CW, Maleimide

Product Code. 30306100
Change view
Click to view available options
:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30306100 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306100 Supplier Tocris Bioscience™ Supplier No. 8880/500U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Near-infrared dye (Reactive group: maleimide)

Key information: IRDye™ 800CW, Maleimide is a near-infrared (NIR) fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity). Suitable for live cell imaging.

Application: Fluorescence microscopy.

Properties and Photophysical Data: Excitation and emission maxima (λ) are 773 nm and 792 nm, respectively.

Please see the product protocol for further information and a guide to protein/antibody labeling.

It is recommended to dissolve this dye in water or DMSO (10 - 20 mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2-((E)-2-((E)-3-(2-((E)-1-(6-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-6-oxohexyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)ethylidene)-2-(4-sulfonatophenoxy)cyclohex-1-en-1-yl)vinyl)-3,3-dimethyl-1-(4-sulfonatobutyl)-3H-indol-1-ium-5
CAS 1279564-25-8
Quantity 500 μg
Target Fluorescent Dyes
Molecular Formula C52H57N4Na3O16S4
Purity 0.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.