missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IRDye™ 680RD, Maleimide

Product Code. 30306124
Click to view available options
:
500 μg
This item is not returnable. View return policy

Product Code. 30306124

Brand: Tocris Bioscience™ 8877/500U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Near-infared dye (reactive group maleimide)

Key information: IRDye™ 680RD, Maleimide is a near-infrared (NIR) fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity).

Application: Fluorescence microscopy.

Properties and Photophysical Data: Excitation and emission maxima (λ) are 680 nm and 694 nm, respectively.

Please see the product protocol for further information and a guide to protein/antibody labeling.

It is recommended to dissolve dye in water or DMSO (10 - 20 mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium (Z)-2-((2Z,4E)-5-(5-Chloro-3,3-dimethyl-7-(3-sulfonatopropyl)-3H-pyrrolo[2,3-b]pyridin-7-ium-2-yl)-3-(3-(3-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-3-oxopropyl)phenyl)penta-2,4-dien-1-ylidene)-3,3-dimethyl-1-(3-sulfonatopropyl)indolin
CAS 1373919-90-4
Quantity 500 μg
Target Fluorescent Dyes
Molecular Formula C45H48ClN5Na2O12S3
Purity 0.8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.