missing translation for 'onlineSavingsMsg'
Learn More

Irbesartan N-Beta-D-Glucuronide, TRC

Product Code. 30389735
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389735 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389735 Supplier Toronto Research Chemicals Supplier No. TRCI7510105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Irbesartan N-beta-D-Glucuronide
CAS 160205-58-3
Molecular Formula C31 H36 N6 O7
Quantity 5 mg
Synonym Irbesartan-Beta-D-Glucuronide
InChI Formula InChI=1S/C31H36N6O7/c1-2-3-10-22-32-31(15-6-7-16-31)30(43)36(22)17-18-11-13-19(14-12-18)20-8-4-5-9-21(20)27-33-34-35-37(27)28-25(40)23(38)24(39)26(44-28)29(41)42/h4-5,8-9,11-14,23-26,28,38-40H,2-3,6-7,10,15-17H2,1H3,(H,41,42)/t23-,24-,25+,26-,28?/m0/s1
SMILES CCCCC1=NC2(CCCC2)C(=O)N1Cc3ccc(cc3)c4ccccc4c5nnnn5C6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O
IUPAC Name (2S,3S,4S,5R)-6-[5-[2-[4-[(2-butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl]phenyl]phenyl]tetrazol-1-yl]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 604.65
Formula Weight 604.2645
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.