Learn More
IRAK4 Mouse anti-Human, Unconjugated, Clone: 1A4C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67180-1-IG-150UL
This item is not returnable.
View return policy
Description
The IRAK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IRAK4. This antibody recognizes human antigen. The IRAK4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IRAK4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1A4C4 | |
| Unconjugated | |
| 150 μL | |
| 51135 | |
| IgG2a |