Learn More
Invitrogen™ IRAK4 Monoclonal Antibody (2H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515883
Description
MA5-15883 targets IRAK4 in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15883 immunogen is purified recombinant fragment of human IRAK4 expressed in E. Coli. . MA5-15883 detects IRAK4 which has a predicted molecular weight of approximately 52kDa.
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
Specifications
| IRAK4 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q8R4K2, Q9NWZ3 | |
| IRAK4 | |
| Purified recombinant fragment of human IRAK4 expressed in E. Coli. | |
| RUO | |
| 266632, 51135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2H9 | |
| Unconjugated | |
| IRAK4 | |
| 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.