missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4-IN-7, MedChemExpress
Description
IRAK4-IN-7 is a selective, potent and orally active interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor, extracted from patent WO2015104688 (example 1). IRAK4-IN-7 has the potential for cancer and inflammatory diseases treatment.
Specifications
Specifications
| Chemical Name or Material | IRAK4-IN-7 |
| CAS | 1801343-74-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19N7O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 13.89 mg/mL (33.28 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC=CC(C2=CC=C(N)N=C2)=N1)NC3=CN=C4C(OC(N5CCOCC5)=N4)=C3 |
| Molecular Weight (g/mol) | 417.42 |
| Formula Weight | 417.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?