missing translation for 'onlineSavingsMsg'
Learn More

IRAK4 Antibody (10H4), Novus Biologicals™

Product Code. 18357365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357365 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357365 Supplier Novus Biologicals Supplier No. NBP315153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK4
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 10H4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry, ELISA
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Host Species Rabbit
Immunogen A synthesized peptide derived from human IRAK4. UniProt ID (Q9NWZ3).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.