missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK1, GST Tag Recombinant Protein
IRAK1 Recombinant Human Protein
£432.00 - £6900.00
Specifications
Accession Number | NP_001560 |
---|---|
Gene Alias | IRAK1, IRAK, pelle |
Species | Human |
Protein Tag | GST Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11515036
|
Invitrogen™
PV4403 |
10μg |
£432.00
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10513143
|
Invitrogen™
PV4439 |
1mg |
£6900.00
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458653
|
Invitrogen™
PR8140B |
100 μg |
£1602.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
IL-1 receptor associated kinases (IRAKs) are serine⁄threonine kinases involved in signaling via Toll⁄IL-1 pathways.Specifications
NP_001560 | |
Human | |
Baculovirus |
IRAK1, IRAK, pelle | |
GST Tag |
For Research Use Only. Not for use in diagnostic procedures.