missing translation for 'onlineSavingsMsg'
Learn More

IRAK1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Código de producto. 18422573 Tienda Bio Techne Productos
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
18422573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18422573 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP177068MFV450

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

IRAK1 Polyclonal antibody specifically detects IRAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen IRAK1
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the carboxy terminus of human IRAK. The immunogen is located within the last 50 amino acids of IRAK. Amino Acid Squence: DRQGPEESDEFQS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3654
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.