missing translation for 'onlineSavingsMsg'
Learn More

IRAK inhibitor 6, MedChemExpress

Product Code. 16435618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16435618 10 mg 10mg
16421787 50 mg 50mg
16408317 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16435618 Supplier Medchem Express Supplier No. HY13280
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IRAK inhibitor 6 is an inhibitor of interleukin-1 receptor associated kinase 4 (IRAK-4) with IC50 of 160 nM.

IRAK inhibitor 6 is an inhibitor of interleukin-1 receptor associated kinase 4 (IRAK-4) with IC50 of 160 nM.

Specifications

Chemical Name or Material IRAK inhibitor 6
CAS 1042672-97-8
Color Brown
Purity Grade Notes Research
Molecular Formula C20H20N4O3S
Quantity 10 mg
Solubility Information DMSO : 8.33 mg/mL (21.01 mM; Need ultrasonic)
SMILES COC1=CC(N2CCOCC2)=CC=C1NC(C3=CSC(C4=CC=CN=C4)=N3)=O
Molecular Weight (g/mol) 396.46
Formula Weight 396.46
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.