missing translation for 'onlineSavingsMsg'
Learn More

IRAK inhibitor 1, MedChemExpress

Product Code. 16491777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491777 50 mg 50mg
16458307 5 mg 5mg
16405618 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491777 Supplier Medchem Express Supplier No. HY13275
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IRAK inhibitor 1 is a potent IRAK-4 inhibitor with IC50 of 216 nM, is poorly active against JNK-1 and JNK-2 with IC50 of 3.801 μM, and >10 μM, respectively.

IRAK inhibitor 1 is a potent IRAK-4 inhibitor with IC50 of 216 nM, is poorly active against JNK-1 and JNK-2 with IC50 of 3.801 μM, and >10 μM, respectively.

Specifications

Chemical Name or Material IRAK inhibitor 1
CAS 1042224-63-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H19N5
Quantity 50 mg
Solubility Information DMSO : 16.67 mg/mL (56.82 mM; Need ultrasonic)
SMILES C1(C2=CN=C3N2C=CC=C3)=CC=CC(NC4CCNCC4)=N1
Molecular Weight (g/mol) 293.37
Formula Weight 293.37
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.