missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals IR-125, pure, laser grade

Product Code. p-149612 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
2 g
Packaging:
Glass bottle
Unit Size:
100mg
2g
500mg
This item is not returnable. View return policy

Product Code. 10321541

Brand: Thermo Scientific Chemicals 412541000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Indocyanine Green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Melting Point 235.0°C
Merck Index 15, 5002
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, dmso, acetonitrile
Infrared Spectrum Authentic
Lambda Max 784nm (in methanol)
Formula Weight 774.97
Grade Pure
Molar Absorption Coefficient 237500L/(mole.cm) (dried sample)
Packaging Glass bottle
Color Brown to Green
Quantity 100 mg
Physical Form Crystalline Powder or Crystals
Chemical Name or Material IR-125
Show More Show Less
compliance-icons
Product Identifier
  • IR-125
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.