missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQSEC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25321-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a guanine nucleotide exchange factor for the ARF family of small GTP-binding proteins. The encoded protein is a component of the postsynaptic density at excitatory synapses, and may play a critical role in cytoskeletal and synaptic organization through the activation of selected ARF substrates including ARF1 and ARF6. Mutations in this gene have been implicated in nonsyndromic X-linked mental retardation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| IQSEC2 | |
| Polyclonal | |
| Unconjugated | |
| IQSEC2 | |
| BRAG1, IQ motif and Sec7 domain 2, IQSEC2, KIAA0522 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23096 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JU85 | |
| IQSEC2 | |
| IQSEC2 Fusion Protein Ag18226 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction