missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IQSEC1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA538019

Product Code. 15534951

  • £434.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A recommended positive control is A-20 cell lysate. At least four isoforms of IQSEC1 are known to exist. IQSEC1 antibody is not expected to cross-react with IQSEC2.

ADP ribosylation factors (Arfs) are small GTP-binding proteins known for their role in vesicular transport, where they nucleate the assembly of coat protein complexes at sites of carrier vesicle formation. IQSEC1 (GEP100/BRAG2), a guanine nucleotide exchange protein for ARF6, belongs to the BRAG family and contains one IQ domain, one PH domain and one SEC7 domain. As a known activator of ARF6, IQSEC1 regulates cell surface levels of Beta1 integrin and its knockdown blocked tumour metastasis. IQSEC1 links EGFR signaling to ARF6 activation and may contribute to the cancer metastasis and malignancy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IQSEC1
Polyclonal
Unconjugated
IQSEC1
ADP-ribosylation factors guanine nucleotide-exchange protein 100; ADP-ribosylation factors guanine nucleotide-exchange protein 2; ARFGEP100; ARF-GEP100; AW561907; BC026481; BRAG2; brefeldin A-resistant ARF-GEF2; brefeldin-resistant Arf-GEF 2 protein; cI-43; D6Ertd349e; GEP100; IQ motif and Sec7 domain 1; IQ motif and SEC7 domain-containing protein 1; Iqsec1; Kiaa0763
Rabbit
Antigen affinity chromatography
RUO
232227, 686590, 9922
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q6DN90, Q8R0S2
IQSEC1
19 amino acid peptide near the amino terminus of human IQSEC1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.