missing translation for 'onlineSavingsMsg'
Learn More

IQSEC1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18424445 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424445 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424445 Supplier Novus Biologicals Supplier No. NBP281958AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IQSEC1 Polyclonal antibody specifically detects IQSEC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQSEC1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factors guanine nucleotide-exchange protein 100, ADP-ribosylation factors guanine nucleotide-exchange protein 2, ARFGEP100, BRAG2, IQ motif and Sec7 domain 1, IQ motif and SEC7 domain-containing protein 1, KIAA0763Brefeldin-resistant Arf-GEF 2 protein
Host Species Rabbit
Immunogen IQSEC1 antibody was raised against a 19 amino acid peptide near the amino terminus of human IQSEC1. The immunogen is located within amino acids 60 - 110 of IQSEC1. Amino Acid Squence: PPGQQQRTRRPKLQHSTSI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9922
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.