Learn More
Thermo Scientific™ IQOQ Qualification Service and Documentation for IntelliStack Incubator Shaker
Shop All Thermo Scientific ProductsDescription
- IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements. Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment and any other accessories are assembled and installed. Configuration information for each system component gets recorded.
- Installation of equipment is NOT included
- OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
- Key tests: Door Ajar, Power Failure, Temperature Alerts
All of this will be performed and delivered by a Thermo Fisher Qualified Engineer equipped with training certificates, temperature mapping data logger calibrated annually with metrology certificates.
Specifications
Specifications
| Product Type | IQ/OQ Service |