Learn More
Unity Lab Services™ IQOQ Incubators, Ovens, Vacuum Oven And Furnaces 3rd Party
Product Code. 30286551
Change view
Click to view available options
For Use With (Equipment):
Ovens, Vacuums, Furnaces, Incubators
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
30286551 Ovens, Vacuums, Furnaces, Incubators Each
1 options
Product Code. 30286551 Supplier Unity Lab Services™ Supplier No. IOQCOVMVS
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installation and operational qualification service of ovens, vacuum ovens, furnaces and incubators not manufactured by Thermo Fisher Scientific. Ensures equipment meets specs and functions properly, key tests like door ajar and temperature.
  • Installation and operational qualification service of ovens, vacuum ovens, furnaces and incubators manufactured by third parties.
  • This service will confirm equipment is operating to defined specifications.
  • Operational qualification includes a test of temperature distribution with 5 points on 1 shelf for vacuum oven and 9 points in the work volume for ovens and incubators.
  • The service includes a test of temperature distribution at one temperature setpoint.
  • NOTE! For the support of Memmert products, validation products must be ordered separately under item code 16344567
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ovens, Vacuums, Furnaces, Incubators
Product Type IQ/OQ Service