Learn More
Invitrogen™ IQGAP2 Monoclonal Antibody (K06332_16C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555949
Description
Sequence of this protein is as follows: EQTVVTLRNP NAVLTLVDDN LAPEYQKELW DAKKKKEENA RLKNSCISEE ERDAYEELLT QAEIQGNINK VNRQAAVDHI NAVIPEGDPE NTLLALKKPE AQLPAVYPFA AAMYQNELFN LQKQNTMNYL AHEELLIAVE MLSAVALLNQ ALESNDLVSV QNQLRSPAIG LNNLDKAYVE RYANTLLSVK LEVLSQGQDN LSWNEIQNCI DMVNAQIQEE NDRVVAVGYI NEAIDEGNPL RTLETLLLPT ANISDVDPAH AQHYQDVLYH AKSQKLGDSE SVSKVLWLDE IQQAVDDANV
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
Specifications
| IQGAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13576 | |
| IQGAP2 | |
| Recombinant human IQGAP2 (immunogen range is 231-530). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| K06332_16C7 | |
| Unconjugated | |
| IQGAP2 | |
| 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10788 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.