missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQGAP1 Recombinant Rabbit Monoclonal Antibody (23GB2020), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552473
This item is not returnable.
View return policy
Description
IQGAP1 Recombinant Monoclonal antibody specifically detects IQGAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| IQGAP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IQGAP1 | |
| AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29875, 361598, 8826 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2020 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P46940, Q9JKF1 | |
| IQGAP1 | |
| A synthesized peptide derived from human IQGAP1 (1-58AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction