Learn More
IQGAP1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-22167-20UL
Description
The IQGAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IQGAP1. This antibody recognizes human, mouse antigen. The IQGAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| IQGAP1 | |
| 1000 μg/mL | |
| HUMORFA01, IQGAP1, KIAA0051, p195, SAR1 | |
| 20 μL | |
| 8826 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |