missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQCH Monoclonal Antibody (4H3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522514
This item is not returnable.
View return policy
Description
Peptide Sequence: LSQLITDHLQ IQRWLFKMDS EFRGNGTAFC DIPSYLKCYK WVLKESSRYG LEDWRKKWAQ EPALVKISEE LAGILAQHAQ PVNEKRFPTW RKFLQTFLSQ.
May play a regulatory role in spermatogenesis.
Specifications
| IQCH | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q86VS3 | |
| IQCH | |
| IQCH (NP_073621, 301 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 4H3 | |
| Unconjugated | |
| IQCH | |
| IQ domain-containing protein H; IQ motif containing H; IQCH; NYDSP5; Testis development protein NYD-SP5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64799 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction