missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555915
This item is not returnable.
View return policy
Description
Immunogen sequence: RTVRTQELGL SEDHIIWDGT LGGPEHSVLD LWRTKPPKGQ APTDRSSRDG TSNEPSHEGQ KKQRTIPWRS KSPEILSSTK AGCTGEEQWR GRPWKTEPPG Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 26%.
The IQCC (IQ Motif Containing C) gene encodes a protein that contains the IQ motif, which is known for its role in binding calmodulin and other proteins with EF-hand calcium-binding motifs. IQCC is located on chromosome 6 at position 6q16.1. The IQCC protein is involved in several cellular processes, including signaling pathways related to intracellular calcium levels and regulation of ion channels. Additionally, recent studies have indicated its potential involvement in cancer biology, where disruptions in its expression or function may contribute to tumorigenesis. The exact molecular mechanisms and pathways influenced by IQCC are still under investigation, and elucidating these aspects could uncover new therapeutic targets for various diseases, including cancer.
Specifications
| IQCC | |
| Polyclonal | |
| Unconjugated | |
| IQCC | |
| BC026978; IQ domain-containing protein C; IQ motif containing C; Iqcc; RP4-622L5.6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55721 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4KMZ1 | |
| IQCC | |
| Recombinant protein corresponding to Human IQCC. Recombinant protein control fragment (Product #RP-94346). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction