missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQCB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112923
This item is not returnable.
View return policy
Description
IQCB1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| IQCB1 | |
| Polyclonal | |
| Unconjugated | |
| Iqcb1 | |
| 6820449I09Rik; AV128382; IQ calmodulin-binding motif containing 1; IQ calmodulin-binding motif-containing protein 1; IQ motif containing B1; IQCB1; KIAA0036; nephrocystin 5; Nephrocystin-5; NPHP5; OK/SW-cl.85; p53 and DNA damage-regulated IQ motif protein; PIQ; SLSN5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 303915, 320299, 9657 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.03 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q15051, Q8BP00 | |
| Iqcb1 | |
| Recombinant Human IQ calmodulin-binding motif-containing protein 1 protein (445-598AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction